missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SEC11A Polyclonal Antibody

Description
Immunogen sequence: SPIVVVLSGSM EPAFHRGDLL FLTN Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.
This gene encodes a member of the peptidase S26B family. The encoded protein is an 18kDa subunit of the signal peptidase complex and has been linked to cell migration and invasion, gastric cancer and lymph node metastasis. Alternative splicing results in multiple transcript variants. A related pseudogene has been identified on chromosome 8.
Specifications
Specifications
| Antigen | SEC11A |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | SEC11A |
| Gene Accession No. | P67812 |
| Gene Alias | 1810012E07Rik; 18kDa; Endopeptidase SP18; microsomal signal peptidase 18 kDa subunit; SEC11 homolog A; SEC11 homolog A (S. cerevisiae); SEC11 homolog A, signal peptidase complex subunit; SEC11A; Sec11l1; Sec11-like 1; SEC11-like protein 1; sid 2895; Sid2895; Sid2895p; signal peptidase complex (18kD); signal peptidase complex 18; signal peptidase complex 18kD; signal peptidase complex catalytic subunit SEC11A; SPase 18 kDa subunit; SPC18; SPCS4A |
| Gene Symbols | SEC11A |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?