missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEC13 Antibody (1280B), Alexa Fluor™ 700, R&D Systems™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals MAB90551AF700
This item is not returnable.
View return policy
Description
SEC13 Monoclonal specifically detects SEC13 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| SEC13 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| SEC13 | |
| rhSEC13 aa123-322 | |
| 100 μL | |
| metabolism | |
| 6396 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| 1280B | |
| Western Blot | |
| D3S1231ESEC13L1npp-20, protein SEC13 homolog, SEC13 (S. cerevisiae)-like 1, SEC13 homolog (S. cerevisiae), SEC13L1, SEC13-like 1 isoform, SEC13-like protein 1, SEC13-related protein, SEC13RSEC13-like 1 (S. cerevisiae) | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction