missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEC14L1 Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97656G
This item is not returnable.
View return policy
Description
SEC14L1 Polyclonal specifically detects SEC14L1 in Human samples. It is validated for Western Blot.
Specifications
| SEC14L1 | |
| Polyclonal | |
| Western Blot | |
| DKFZp686C06176, PRELID4A, SEC14L, SEC14-like 1 (S. cerevisiae), SEC14-like protein 1 | |
| Produced in rabbits immunized with E. coli-derived Human SEC14L1 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 6397 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction