missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEC14L2 Monoclonal Antibody (OTI1H3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503561
This item is not returnable.
View return policy
Description
This gene encodes a cytosolic protein which belongs to a family of lipid-binding proteins including Sec14p, alpha-tocopherol transfer protein, and cellular retinol-binding protein. The encoded protein stimulates squalene monooxygenase which is a downstream enzyme in the cholesterol biosynthetic pathway. Alternatively spliced transcript variants encoding different isoforms have been identified for this gene.Specifications
| SEC14L2 | |
| Monoclonal | |
| 0.59 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O76054 | |
| SEC14L2 | |
| Full length human recombit protein of human SEC14L2 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI1H3 | |
| Unconjugated | |
| SEC14L2 | |
| C22orf6, SPF, TAP, TAP1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 23541 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction