missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEC14L2 Monoclonal Antibody (OTI1H3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503561S
This item is not returnable.
View return policy
Description
SEC14L2 Monoclonal antibody specifically detects SEC14L2 in Human samples. It is validated for Immunocytochemistry, Western BlotSpecifications
| SEC14L2 | |
| Monoclonal | |
| 0.59 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P41970 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 2004 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Immunocytochemistry, Western Blot | |
| OTI1H3 | |
| Unconjugated | |
| ELK3 | |
| 1300013M05Rik; Alpha-tocopherol-associated protein; C22orf6; KIAA1186; KIAA1658; SEC14 like lipid binding 2; SEC14L2; SEC14-like 2; SEC14-like 2 (S. cerevisiae); SEC14-like lipid binding 2; SEC14-like protein 2; SEC14-like protein 2-like; SPF; squalene transfer protein; Supernatant protein factor; TAP; TAP1; tocopherol-associated protein; tocopherol-associated protein 1 | |
| Full length human recombinant protein of human ELK3 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction