Learn More
Invitrogen™ SEC14L2 Monoclonal Antibody (OTI4H2)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA525728
Product Code. 15985324
  • € 572.00 / 100µL
Description
SEC14L2 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
This gene encodes a cytosolic protein which belongs to a family of lipid-binding proteins including Sec14p, alpha-tocopherol transfer protein, and cellular retinol-binding protein. The encoded protein stimulates squalene monooxygenase which is a downstream enzyme in the cholesterol biosynthetic pathway. Alternatively spliced transcript variants encoding different isoforms have been identified for this gene.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SEC14L2
Monoclonal
0.36 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
O76054, Q99J08
SEC14L2
Full length human recombinant protein of SEC14L2 produced in HEK293T cell.
100 μL
Primary
Human, Mouse, Canine, Monkey
Antibody
IgG1
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
OTI4H2
Unconjugated
SEC14L2
1300013M05Rik; Alpha-tocopherol-associated protein; C22orf6; KIAA1186; KIAA1658; SEC14 like lipid binding 2; SEC14L2; SEC14-like 2; SEC14-like 2 (S. cerevisiae); SEC14-like lipid binding 2; SEC14-like protein 2; SEC14-like protein 2-like; SPF; squalene transfer protein; Supernatant protein factor; TAP; TAP1; tocopherol-associated protein; tocopherol-associated protein 1
Mouse
Affinity Chromatography
RUO
23541, 477539, 67815
-20°C, Avoid Freeze/Thaw Cycles