missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SEC14L3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113471
This item is not returnable.
View return policy
Description
SEC14L3 Polyclonal Antibody for Western Blot, IHC (P), ELISA
The protein encoded by this gene is highly similar to the protein encoded by the Saccharomyces cerevisiae SEC14 gene. The SEC14 protein is a phophatidylinositol transfer protein that is essential for biogenesis of Golgi-derived transport vesicles, and thus is required for the export of yeast secretory proteins from the Golgi complex. The specific function of this protein has not yet been determined. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
Specifications
| SEC14L3 | |
| Polyclonal | |
| Unconjugated | |
| SEC14L3 | |
| 1110069O07Rik; 45 kDa secretory protein; rsec45; SEC14 like lipid binding 3; Sec14l3; SEC14-like 3; SEC14-like 3 (S. cerevisiae); SEC14-like lipid binding 3; SEC14-like protein 3; SEC14p-like protein TAP2; secretory protein, 45kD; Spf2; TAP2; Tocopherol-associated protein 2 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 266629, 380683 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 3 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q9UDX4 | |
| SEC14L3 | |
| Recombinant Human SEC14-like protein 3 protein (201-400AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction