missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SEC22A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144171
This item is not returnable.
View return policy
Description
SEC22A Polyclonal Antibody for ICC/IF, IHC (P), ELISA
The protein encoded by this gene belongs to the member of the SEC22 family of vesicle trafficking proteins. This protein has similarity to rat SEC22 and may act in the early stages of the secretory pathway. [provided by RefSeq, Nov 2008].
Specifications
| SEC22A | |
| Polyclonal | |
| Unconjugated | |
| SEC22A | |
| SEC22 vesicle-trafficking protein homolog A; SEC22 vesicle-trafficking protein-like 2; SEC22A; SEC22L2; Vesicle-trafficking protein SEC22a | |
| Rabbit | |
| Protein G | |
| RUO | |
| 26984 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 3 mg/mL | |
| PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
| Q96IW7 | |
| SEC22A | |
| Recombinant Human Vesicle-trafficking protein SEC22a protein (10-187AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction