missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEC23A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
€ 218.00
Specifications
| Antigen | SEC23A |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 2 μg/mL, Flow Cytometry 3 - 5 ug 106 cells, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:200, Immunoprecipitation 1:10 - 1:500 |
| Applications | Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
| Classification | Polyclonal |
Description
SEC23A Polyclonal specifically detects SEC23A in Human, Mouse, Rat, Hamster samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
| SEC23A | |
| Western Blot 2 μg/mL, Flow Cytometry 3 - 5 ug 106 cells, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:200, Immunoprecipitation 1:10 - 1:500 | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse, Rat, Hamster | |
| B5DFC3 | |
| 10484 | |
| Synthetic peptide corresponding to residues M(1) T T Y L E F I Q Q N E E R D G V R (18) C of rat COPII. | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| RUO | |
| PBS and 1 mg/mL BSA with 0.05% Sodium Azide | |
| CLSD, MGC26267, protein transport protein Sec23A, Sec23 (S. cerevisiae) homolog A, Sec23 homolog A (S. cerevisiae), SEC23-related protein A | |
| SEC23A | |
| IgG | |
| Affinity Purified | |
| Cross-reacts with Human, Rat and Mouse. Reported reactivity in Drosopholia. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title