Learn More
Invitrogen™ SEC23B Monoclonal Antibody (OTI6E8)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA527258
Product Code. 15996844
  • € 572.00 / 100µL
Description
SEC23B Monoclonal Antibody for Western Blot
The protein encoded by this gene is a member of the SEC23 subfamily of the SEC23/SEC24 family, which is involved in vesicle trafficking. The encoded protein has similarity to yeast Sec23p component of COPII. COPII is the coat protein complex responsible for vesicle budding from the ER. The function of this gene product has been implicated in cargo selection and concentration. Multiple alternatively spliced transcript variants encoding the same protein have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SEC23B
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q15437
SEC23B
Human recombinant protein fragment corresponding to amino acids 392-641 of SEC23B produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Western Blot
OTI6E8
Unconjugated
SEC23B
CDAII; CDA-II; CDAN2; CWS7; HEMPAS; hSec23B; Protein transport protein Sec23B; Sec23 homolog B; Sec23 homolog B (S. cerevisiae); Sec23 homolog B, coat complex II component; Sec23 homolog B, COPII coat complex component; Sec23b; SEC23B (S. cerevisiae); SEC23-like protein B; SEC23-related protein B; transport protein SEC23B
Mouse
Affinity Chromatography
RUO
10483
-20°C, Avoid Freeze/Thaw Cycles