missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SEC23IP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558378
This item is not returnable.
View return policy
Description
Immunogen sequence: HLELKESLSR MGSDLKQGFI SSLKSAWQTL NEFARAHTSS TQLQEELEKV ANQIKEEEEK QVVEAEKVVE SPDFSKDEDY LGKVGML Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 98%.
SEC23IP is involved in protein transport from the endoplasmic reticulum to the Golgi apparatus. The protein encoded by this gene was identified by its interaction with a mouse protein similar to yeast Sec23p, an essential component of the COPII. This protein shares significant similarity with phospholipid-modifying proteins, especially phosphatidic acid preferring-phospholipase A1. Overexpression of this protein has been shown to cause disorganization of the endoplasmic reticulum-Golgi intermediate compartment and Golgi apparatus, which suggests its role in the early secretory pathway.
Specifications
| SEC23IP | |
| Polyclonal | |
| Unconjugated | |
| SEC23IP | |
| D7Ertd373e; MSTP053; P125; P125A; SEC23 interacting protein; SEC23-interacting protein; Sec23-interacting protein p125; SEC23-interacting protein-like protein; SEC23IP | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 11196, 207352, 309010 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.10 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6NZC7, Q9Y6Y8 | |
| SEC23IP | |
| Recombinant protein corresponding to Human SEC23IP. Recombinant protein control fragment (Product #RP-97033). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction