Learn More
SEC24D Antibody (S01-5G5), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-19915-100ul
 View more versions of this product
Product Code. 18647774
  • € 645.00 / 100µL
Description
SEC24D Monoclonal antibody specifically detects SEC24D in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SEC24D
Monoclonal
0.3 mg/mL
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Rabbit
Affinity purified
RUO
9871
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Western Blot
S01-5G5
Unconjugated
FLJ43974, KIAA0755member D, SEC24 family, member D (S. cerevisiae), SEC24 related gene family, member D
A synthetic peptide of human SEC24D (Uniprot # O94855)
100 μg
Primary
Human, Mouse, Rat