missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SEC24D Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559651
This item is not returnable.
View return policy
Description
Immunogen sequence: ASQLILPDSM KVLPVYMNCL LKNCVLLSRP EISTDERAYQ RQLVMTMGVA DSQLFFYPQL LPIHTLDVKS TMLPAAVRCS ESRLS Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 93%.
The protein encoded by this gene is a member of the SEC24 subfamily of the SEC23/SEC24 family, which is involved in vesicle trafficking. The encoded protein has similarity to yeast Sec24p component of COPII. COPII is the coat protein complex responsible for vesicle budding from the ER. This gene product is implicated in the shaping of the vesicle, and also in cargo selection and concentration.
Specifications
| SEC24D | |
| Polyclonal | |
| Unconjugated | |
| SEC24D | |
| 2310020L09Rik; CLCRP2; Gm1349; KIAA0755; Protein transport protein Sec24D; SEC24 family member D; SEC24 family, member D; SEC24 homolog D, COPII coat complex component; SEC24 related gene family, member D; Sec24 related gene family, member D (S. cerevisiae); SEC24D; SEC24-related protein D | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9871 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O94855 | |
| SEC24D | |
| Recombinant protein corresponding to Human SEC24D. Recombinant protein control fragment (Product #RP-96685). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction