missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SEC24D Recombinant Rabbit Monoclonal Antibody (6F8S4)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA555552
This item is not returnable.
View return policy
Description
Immunogen Sequence: MSQQGYVATP PYSQPQPGIG LSPPHYGHYG DPSHTASPTG MMKPAGPLGA TATRGMLPPG PPPPGPHQFG QNGAHATGHP PQRFPGPPPV NNVASSHAPY.
The protein encoded by this gene is a member of the SEC24 subfamily of the SEC23/SEC24 family, which is involved in vesicle trafficking. The encoded protein has similarity to yeast Sec24p component of COPII. COPII is the coat protein complex responsible for vesicle budding from the ER. This gene product is implicated in the shaping of the vesicle, and also in cargo selection and concentration.
Specifications
| SEC24D | |
| Recombinant Monoclonal | |
| 1.2 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O94855 | |
| SEC24D | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-100 of human SEC24D. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 6F8S4 | |
| Unconjugated | |
| SEC24D | |
| 2310020L09Rik; CLCRP2; Gm1349; KIAA0755; Protein transport protein Sec24D; SEC24 family member D; SEC24 family, member D; SEC24 homolog D, COPII coat complex component; SEC24 related gene family, member D; Sec24 related gene family, member D (S. cerevisiae); SEC24D; SEC24-related protein D | |
| Rabbit | |
| Protein A | |
| RUO | |
| 310843, 69608, 9871 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction