missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEC3 Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-78752F
This item is not returnable.
View return policy
Description
SEC3 Polyclonal antibody specifically detects SEC3 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| SEC3 | |
| Polyclonal | |
| Western Blot, Immunoprecipitation | |
| BM-102, exocyst complex component 1, Exocyst complex component Sec3, FLJ10893, SEC3L1, SEC3-like 1, SEC3-like 1 (S. cerevisiae), SEC3P, SEC3Sec3p | |
| The immunogen for this product maps to a region between residue 100 and 150 of human Exocyst Complex Component Sec3 using the numbering given in entry NP_060731.2 (GeneID 55763). | |
| 0.1 mL | |
| Signal Transduction | |
| 55763 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| FITC | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction