missing translation for 'onlineSavingsMsg'
Learn More

SEC31A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16817924 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16817924 20 μL 20µL
16807924 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16817924 Supplier Proteintech Supplier No. 179131AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The protein encoded by this gene is similar to yeast Sec31 protein. Yeast Sec31 protein is known to be a component of the COPII protein complex which is responsible for vesicle budding from endoplasmic reticulum (ER). This protein was found to colocalize with SEC13, one of the other components of COPII, in the subcellular structures corresponding to the vesicle transport function. An immunodepletion experiment confirmed that this protein is required for ER-Golgi transport. Alternative splicing results in multiple transcript variants encoding different isoforms.

Specifications

Antigen SEC31A
Applications Immunofluorescence, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SEC31A
Gene Accession No. O94979, Q3UPL0, Q9Z2Q1
Gene Alias ABP125, ABP130, HSPC275, HSPC334, KIAA0905, SEC31 like protein 1, SEC31 related protein A, SEC31A, SEC31L1, Web1 like protein
Gene Symbols Sec31a
Host Species Rabbit
Immunogen SEC31A Fusion Protein Ag12314
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22872, 69162, 93646
Target Species Human, Rat, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.