missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SEC61alpha Recombinant Rabbit Monoclonal Antibody (24GB1230)
GREENER_CHOICE

Product Code. p-200111063
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30276088 100 μL 100µL
30275617 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30276088 Supplier Invitrogen™ Supplier No. MA557247

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

SEC61alpha Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

The SEC61 complex is a major component of the endoplasmic reticulum (ER) translocation site. This complex is composed of three subunits: SEC61alpha, SEC61beta and SEC61gamma. SEC61alpha is a polytopic integral membrane protein. It is a core component of the ER translocation complex, playing a crucial role in the insertion of secretory and membrane polypeptides into the ER.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SEC61alpha
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 24GB1230
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SEC61A2
Gene Accession No. Q9H9S3, Q9JLR1
Gene Alias protein transport protein Sec61 subunit alpha; protein transport protein Sec61 subunit alpha isoform 2; Sec61 alpha; Sec61 alpha 2 subunit; Sec61 alpha 2 subunit (S. cerevisiae); Sec61 alpha form 2; sec61 alpha-2; Sec61 translocon alpha 2 subunit; Sec61, alpha subunit 2; Sec61, alpha subunit 2 (S. cerevisiae); SEC61A2
Gene Symbols SEC61A2
Host Species Rabbit
Immunogen A synthesized peptide derived from human SEC61A1/2 (aa 350-450).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 361273, 55176, 57743
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.