missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEC63 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13978-1-AP-150UL
This item is not returnable.
View return policy
Description
SEC63 gene ontology annotations related to this gene include liver development; multicellular organism aging; nitrogen compound metabolic process; posttranslational protein targeting to endoplasmic reticulum membrane; posttranslational protein targeting to membrane, translocation; renal system development; SRP-dependent cotranslational protein targeting to membrane.Specifications
| SEC63 | |
| Polyclonal | |
| Unconjugated | |
| SEC63 | |
| ERdj2, PRO2507, SEC63, SEC63 homolog (S. cerevisiae), SEC63L | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11231, 140740, 309858 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8VHE0, Q9UGP8 | |
| Sec63 | |
| SEC63 Fusion Protein Ag5021 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction