missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Sec8 Monoclonal specifically detects Sec8 in Human, Mouse, Rat, Bovine, Canine, Chicken, Fish, Guinea Pig, Hamster, Primate, Rabbit, Sheep samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | Sec8 |
| Applications | Western Blot, Immunohistochemistry, Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 14G1 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 12.5 ug/ml, Immunohistochemistry-Paraffin 1:10-1:500 |
| Formulation | PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide |
| Gene Alias | exocyst complex component 4, KIAA1699SEC8L1, MGC27170, REC8, SEC8Exocyst complex component Sec8, SEC8-like 1, SEC8-like 1 (S. cerevisiae), Sec8p |
| Gene Symbols | EXOC4 |
| Host Species | Mouse |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?