missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Secernin-1 Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77187R
This item is not returnable.
View return policy
Description
Secernin-1 Polyclonal antibody specifically detects Secernin-1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Secernin-1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 550 | |
| KIAA0193SES1, secernin 1, secernin-1 | |
| Antibody was raised against a 20 amino acid synthetic peptide near the carboxy terminus of human SCRN1. The immunogen is located within amino acids 320 - 370 of SCRN1. Amino Acid Squence: CPAKKEPRFQEKPDRRHELYK | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication | |
| 9805 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction