missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SECISBP2 Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-22301G
This item is not returnable.
View return policy
Description
SECISBP2 Polyclonal antibody specifically detects SECISBP2 in Human samples. It is validated for Western Blot
Specifications
| SECISBP2 | |
| Polyclonal | |
| Western Blot | |
| DKFZp686C09169, SBP2SECIS-binding protein 2, SECIS binding protein 2, selenocysteine insertion sequence binding protein 2, selenocysteine insertion sequence-binding protein 2 | |
| The immunogen this antibody was made to, maps to a region between residue 804 to 854 of human SECIS Binding Protein 2 using the numbering given in entry NP_076982.3 (GeneID 79048). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 79048 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction