Learn More
SECISBP2L Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP3-05914MFV450
Product Code. 18505152
  • € 428.00 / 0.10mL
Description
SECISBP2L Polyclonal antibody specifically detects SECISBP2L in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polyclonal
Produced in rabbits immunized with E. coli-derived Human SECISBP2L fragment.
Store at 4C in the dark.
KIAA0256SECIS-binding protein 2-like, SECIS binding protein 2-like, selenocysteine insertion sequence-binding protein 2-like
0.1 mL