Learn More
3M™ SecureFit™ Anti-Scratch, Anti-Fog Safety Spectacles
8.20 EUR valid until 2024-12-31
Use promo code "22912" to get your promotional price.
8.20 EUR valid until 2024-04-30
Use promo code "21673" to get your promotional price.
Use promo code "22912" to get your promotional price.
8.20 EUR valid until 2024-04-30
Use promo code "21673" to get your promotional price.
Click to view available options
Lens Tint:
Amber
Blue Mirror
Clear
Grey
Indoor/Outdoor
Description
- Available in a range of lens tints
- Protection against UV radiation
- Naturally adjusts to individuals' head sizes
- Rimless lenses and fixed-length temple arm to enhance frame comfort
- Soft, adjustable nose pads for even greater personalization of fit
- Get added comfort over the ears with padded temple touchpoints
- Temple arms in a bold colour for an infusion of style
Specifications
Lens Tint | Indoor/Outdoor |
Product Type | Goggles |
No. per Pack | 1 Ea. |
Lens Material | Polycarbonate |