missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SEL1L Polyclonal Antibody

Product Code. 17177243
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17177243 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17177243 Supplier Invitrogen™ Supplier No. PA5109312

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Control: Zebrafish.

The protein encoded by this gene is part of a protein complex required for the retrotranslocation or dislocation of misfolded proteins from the endoplasmic reticulum lumen to the cytosol, where they are degraded by the proteasome in a ubiquitin-dependent manner. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SEL1L
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.2% BSA and 0.05% sodium azide; pH 7.4
Gene SEL1L
Gene Accession No. 0
Gene Alias AW493766; fj60e07; IBD2; mKIAA4137; PRO1063; Protein sel-1 homolog 1; Sel1 (suppressor of lin-12) 1 homolog; sel-1 suppressor of lin-12-like; sel-1 suppressor of lin-12-like (C. elegans); sel-1 suppressor of lin-12-like 1; Sel1h; SEL1L; sel-1L; SEL1L adaptor subunit of ERAD E3 ubiquitin ligase; SEL1L ERAD E3 ligase adaptor subunit; SEL1L, ERAD E3 ligase adaptor subunit; SEL1L1; SEL1-LIKE; si:dkey-177p2.3; Suppressor of lin 12 (sel-1), C. elegans, homolog of; suppressor of lin-12-like protein 1; TSA305; UNQ128/PRO1063; wu:fj60e07; zgc:56075
Gene Symbols SEL1L
Host Species Rabbit
Immunogen Recombinant protein within zebrafish SEL1L aa 109-281.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 568615
Target Species Zebrafish
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.