missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEL1L3 Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97792MFV610
This item is not returnable.
View return policy
Description
SEL1L3 Polyclonal antibody specifically detects SEL1L3 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| SEL1L3 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin | |
| DKFZp781J1697, FLJ21629, KIAA0746FLJ41299, protein sel-1 homolog 3, sel-1 suppressor of lin-12-like 3 (C. elegans), Sel-1L3, Suppressor of lin-12-like protein 3 | |
| Produced in rabbits immunized with E. coli-derived Human SEL1L3 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 23231 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction