missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SEL1L3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15736911 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15736911 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15736911 Supplier Invitrogen™ Supplier No. PA559042

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: SMNLGYKHYQ GIDNYPLDWE LSYAYYSNIA TKTPLDQHTL QGDQAYVETI RLKDDEILKV QTKEDGDVFM WLKHEATRGN AAAQQRLAQM LFWGQQGVAK NPEAA Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.

SEL1L3 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SEL1L3
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SEL1L3
Gene Accession No. Q68CR1
Gene Alias KIAA0746; Protein sel-1 homolog 3; sel-1 suppressor of lin-12-like 3; SEL1L family member 3; SEL1L3; Sel-1L3; Suppressor of lin-12-like protein 3
Gene Symbols SEL1L3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SEL1L3. Recombinant protein control fragment (Product #RP-98346).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23231
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.