missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Seladin 1 Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98505MFV450
This item is not returnable.
View return policy
Description
Seladin 1 Polyclonal antibody specifically detects Seladin 1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Seladin 1 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 1718 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| mFluor Violet 450 SE | |
| 24-dehydrocholesterol reductaseNbla03646, DCE, desmosterol-to-cholesterol enzyme, Diminuto/dwarf1 homolog, EC 1.3.1.72, KIAA0018SELADIN1, seladin 1,3-beta-hydroxysterol delta-24-reductase, seladin-1, selective AD indicator 1,3 beta-hydroxysterol delta 24-reductase | |
| Produced in rabbits immunized with E. coli-derived Human Seladin 1 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction