missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SELE, Mouse anti-Human, PE, Clone: TEA2/1, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB15349.100Reactions
This item is not returnable.
View return policy
Specifications
| SELE | |
| Monoclonal | |
| PE | |
| In PBS, pH 7.4 (protein stabilizer, 0.09% sodium azide). | |
| CD62E/ELAM/ELAM1/ESEL/LECAM2 | |
| Mouse | |
| 100 Reactions | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Flow Cytometry | |
| TEA2/1 | |
| Flow Cytometry (20 uL/106 cells) The optimal working dilution should be determined by the end user. | |
| selectin E | |
| SELE | |
| Recombinant protein corresponding to full length human SELE. | |
| RUO | |
| 6401 | |
| Store in the dark at 4°C. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction