missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SELENBP1 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-97662CL1
This item is not returnable.
View return policy
Description
SELENBP1 Polyclonal antibody specifically detects SELENBP1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| SELENBP1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| CoraFluor 1 | |
| 56 kDa selenium-binding protein, hSBP, hSP56, LPSB, SBP, SBP56, selenium binding protein 1, selenium-binding protein 1, SP56FLJ13813 | |
| Produced in rabbits immunized with E. coli-derived Human SELENBP1 fragment. | |
| 0.1 mL | |
| Cancer, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
| 8991 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction