missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SELENBP1 Antibody (OTI3H1), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74053AF532
This item is not returnable.
View return policy
Description
SELENBP1 Monoclonal specifically detects SELENBP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| SELENBP1 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| SELENBP1 | |
| Full length recombinant protein of human SELENBP1 (NP_003935) produced in HEK293T cells. | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a |
| Western Blot, Immunohistochemistry | |
| OTI3H1 | |
| Western Blot, Immunohistochemistry | |
| 56 kDa selenium-binding protein, hSBP, hSP56, LPSB, SBP, SBP56, selenium binding protein 1, selenium-binding protein 1, SP56FLJ13813 | |
| Mouse | |
| 0.1 mL | |
| 8991 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction