missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SELENBP1 Monoclonal Antibody (OTI3C6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA504739
This item is not returnable.
View return policy
Description
Selenium is an essential trace element that confers tolerance to toxicity arising through exposure to heavy metals or other reactive xenobiotics. Selenium exhibits potent anticarcinogenic properties, and deficiency of selenium may cause certain neurologic diseases. Both effects are attributed to selenium-binding proteins. Selenium binding protein 1 is down-regulated in lung adenocarcinoma, colorectal cander and ovarian cancer. It is two-fold upregulated in the brains of patients suffering from schizophrenia, and is therefore a biomarker for this disease.Specifications
| SELENBP1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q13228 | |
| SELENBP1 | |
| Full length human recombit protein of human SELENB1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3C6 | |
| Unconjugated | |
| SELENBP1 | |
| HEL-S-134P, LPSB, SBP56, SP56, hSBP | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 8991 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction