Learn More
SELENBP1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 27449-1-AP-150UL
 View more versions of this product
Product Code. 16855625
  • € 410.00 / 150µL
Description
Selenium is an essential trace element that confers tolerance to toxicity arising through exposure to heavy metals or other reactive xenobiotics. Selenium exhibits potent anticarcinogenic properties, and deficiency of selenium may cause certain neurologic diseases. Both effects are attributed to selenium-binding proteins. Selenium binding protein 1 is down-regulated in lung adenocarcinoma, colorectal cander and ovarian cancer. It is two-fold upregulated in the brains of patients suffering from schizophrenia, and is therefore a biomarker for this disease.
Specifications
Specifications
SELENBP1
Polyclonal
Unconjugated
SELENBP1
hSBP, hSP56, LPSB, SBP, SBP56, SELENBP1, selenium binding protein 1, SP56
Rabbit
Antigen Affinity Chromatography
RUO
20341, 8991
-20°C
Liquid
Western Blot
0.3 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P17563, Q13228
SELENBP1
SELENBP1 Fusion Protein Ag24453
150 μL
Primary
Human, Mouse