missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Seli Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5139972
This item is not returnable.
View return policy
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: MAYFDPDFYA SAPGHKHVPD WV Highest antigen sequence indentity to the following orthologs: Rat - 91%, Mouse - 91%.
The multi-pass transmembrane protein encoded by this gene belongs to the CDP-alcohol phosphatidyltransferase class-I family. It catalyzes the transfer of phosphoethanolamine from CDP-ethanolamine to diacylglycerol to produce phosphatidylethanolamine, which is involved in the formation and maintenance of vesicular membranes, regulation of lipid metabolism, and protein folding. This protein is a selenoprotein, containing the rare selenocysteine (Sec) amino acid at its active site. Sec is encoded by the UGA codon, which normally signals translation termination. The 3' UTRs of selenoprotein mRNAs contain a conserved stem-loop structure, designated the Sec insertion sequence (SECIS) element, that is necessary for the recognition of UGA as a Sec codon rather than as a stop signal. A pseudogene of this locus has been identified on chromosome 7.
Specifications
| Seli | |
| Polyclonal | |
| Unconjugated | |
| SELENOI | |
| EPT1; ethanolaminephosphotransferase 1; hEPT1; KIAA1724; RGD1560938; SELENOI; Selenoprotein I; SELI; SEPI | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 85465 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9C0D9 | |
| SELENOI | |
| Recombinant protein corresponding to Human SELENOI. Recombinant protein control fragment (Product #RP-109057). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction