Learn More
Keystone Cleanroom Products Self-Sealing 1421B Tyvek™ Autoclave Bag with Steam Indicator, 0 to 12 in. Wide
GREENER_CHOICE
Product Code. p-200049730 Shop All Keystone Cleanroom Products Products
Click to view available options
Dimensions (L x W):
9.5 x 12 in.
4 x 4 in.
7 x 7 in.
8 x 6 in.
9 x 4 in.
9 x 7 in.
10 x 6 in.
11 x 8 in.
12 x 6 in.
13 x 7 in.
15 x 6 in.
15 x 8 in.
16 x 4 in.
16 x 6 in.
20 x 8 in.
26 x 4 in.
28 x 4 in.
30 x 4 in.
38 x 6 in.
12 x 12 in.
14 x 12 in.
15 x 10 in.
16 x 12 in.
18 x 10 in.
18 x 12 in.
20 x 10 in.
20 x 12 in.
22 x 10 in.
22 x 12 in.
24 x 10 in.
24 x 12 in.
28 x 12 in.
30 x 10 in.
30 x 12 in.
32 x 12 in.
34 x 12 in.
36 x 12 in.
40 x 12 in.
42 x 12 in.
46 x 12 in.
48 x 10 in.
48 x 12 in.
56 x 10 in.
62 x 10 in.
Unit Size:
Pack of 250
Pack of 500
Product Code. 30020166
Brand: Keystone Cleanroom Products 6STK0404SS
Only null left
Add to basket
Add to basket
The Self-Sealing 1421B Tyvek™ Autoclave Bag with Steam Indicator is designed for the steam sterilization of pharmaceutical equipment change parts. These bags are closed via the convenient and robust self-sealing closure system.
  • Designed for steam sterilization
  • Tyvek™ on both sides for maximum breathability during sterilization
  • Integrated steam indicator
  • Recommended for sterilization of pharmaceutical equipment
  • Manufactured and packaged in controlled cleanroom environment
  • Eliminates need to cut material and the use of autoclave tape
  • Self-sealing closure is fast and consistent for equipment preparation area
  • Creates secondary level of protection during aseptic presentation of equipment
  • Resists tearing and punctures
  • Superior microbial barrier
  • Certificates of conformance sent with shipment
Specifications
Autoclavable Autoclavable
Capacity (English) 12 oz.
Capacity (Metric) 355 mL
Certifications/Compliance Certificate of Conformance (COC)
Closure Type Self-seal
Color White
Disposable Single-use
For Use With (Application) Steam Sterilization
For Use With (Equipment) Pharmaceutical Equipment and Components
Includes Steam Sterilization Indicator
Material Tyvek
Sterility Non-sterile
Thickness (Metric) 0.13 mm
Temperature Range (English) Up to 250°F
Temperature Range (Metric) Up to 125°C
Thickness (English) 5.3 mil
Dimensions (L x W) 4 x 4 in.
Length (English) 4 in.
Length (Metric) 10 cm
Width (English) 4 in.
Width (Metric) 10 cm
Product Type Autoclave Bag