Learn More
Keystone Cleanroom Products Self-Sealing 1421B Tyvek™ Autoclave Bag with Steam Indicator, 24 to 62 in. Wide
GREENER_CHOICE
Product Code. p-200049732 Shop All Keystone Cleanroom Products Products
Click to view available options
Dimensions (L x W):
12 x 34 in.
16 x 30 in.
16 x 44 in.
20 x 30 in.
24 x 24 in.
24 x 36 in.
24 x 40 in.
28 x 26 in.
28 x 28 in.
28 x 30 in.
28 x 36 in.
30 x 24 in.
30 x 30 in.
32 x 28 in.
32 x 32 in.
34 x 26 in.
34 x 28 in.
34 x 34 in.
36 x 24 in.
36 x 30 in.
36 x 36 in.
38 x 38 in.
40 x 24 in.
40 x 40 in.
42 x 36 in.
42 x 42 in.
44 x 28 in.
44 x 42 in.
46 x 26 in.
46 x 36 in.
48 x 24 in.
48 x 32 in.
48 x 36 in.
48 x 40 in.
48 x 48 in.
50 x 44 in.
50 x 48 in.
50 x 62 in.
52 x 24 in.
54 x 50 in.
56 x 34 in.
60 x 30 in.
60 x 36 in.
60 x 40 in.
62 x 24 in.
64 x 36 in.
64 x 48 in.
72 x 36 in.
72 x 40 in.
Unit Size:
Pack of 100
Pack of 125
Pack of 250
Product Code. 30020015
Brand: Keystone Cleanroom Products 6STK2452SS
Only null left
Add to basket
Add to basket
The Self-Sealing 1421B Tyvek™ Autoclave Bag with Steam Indicator is designed for the steam sterilization of pharmaceutical equipment change parts. These bags are closed via the convenient and robust self-sealing closure system.
  • Designed for steam sterilization
  • Tyvek™ on both sides for maximum breathability during sterilization
  • Integrated steam indicator
  • Recommended for sterilization of pharmaceutical equipment
  • Manufactured and packaged in controlled cleanroom environment
  • Eliminates need to cut material and the use of autoclave tape
  • Self-sealing closure is fast and consistent for equipment preparation area
  • Creates secondary level of protection during aseptic presentation of equipment
  • Resists tearing and punctures
  • Superior microbial barrier
  • Certificates of conformance sent with shipment
Specifications
Autoclavable Autoclavable
Capacity (English) 8.97 gal.
Capacity (Metric) 34 L
Certifications/Compliance Certificate of Conformance (COC)
Closure Type Self-seal
Color White
Disposable Single-use
For Use With (Application) Steam Sterilization
For Use With (Equipment) Pharmaceutical Equipment and Components
Includes Steam Sterilization Indicator
Material Tyvek
Sterility Non-sterile
Thickness (Metric) 0.13 mm
Temperature Range (English) Up to 250°F
Temperature Range (Metric) Up to 125°C
Thickness (English) 5.3 mil
Dimensions (L x W) 52 x 24 in.
Length (English) 52 in.
Length (Metric) 132 cm
Width (English) 24 in.
Width (Metric) 61 cm
Product Type Autoclave Bag