Learn More
Kartell™ Self-Sprung Yellow Forceps
missing translation for 'orderingAttributeHoverText'
Length (Metric):
118 mm
253 mm
missing translation for 'unitSize'
Pack of 5
Description
- Autoclavable
- Yelow
- Self-spring with rounded ends
- Constructed of POM
Specifications
Specifications
| No. per Pack | 5/Pack |
| Material | POM |
| Length (Metric) | 118 mm |
| Product Type | Forceps |
| Type | Forceps |
| Description | Yellow Forceps |