missing translation for 'onlineSavingsMsg'
Learn More

Selitrectinib, MedChemExpress

Product Code. p-200099510
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16492816

Brand: Medchem Express HY101977

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Selitrectinib (LOXO-195) is a next-generation TRK kinase inhibitor, with IC50s of 0.6 nM and TRKA and TRKC, respectively.

Selitrectinib (LOXO-195) is a next-generation TRK kinase inhibitor, with IC50s of 0.6 nM and TRKA and TRKC, respectively.

Specifications

Chemical Name or Material Selitrectinib
Physical Form Solid
CAS 2097002-61-2
Color Off-White
Quantity 25 mg
Purity Grade Notes Research
Molecular Formula C20H21FN6O
For Use With (Application) Cancer-Kinase/protease
Synonym LOXO-195
Solubility Information DMSO : 62.5 mg/mL (164.29 mM; Need ultrasonic)
SMILES FC1=CN=C(CC[C@H](N2)C)C([C@](CCC3)([H])N3C(C=CN4N=C5)=NC4=C5C2=O)=C1
Molecular Weight (g/mol) 380.42
Formula Weight 380.42
Percent Purity 98.06%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.