missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SELK Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-26091G
This item is not returnable.
View return policy
Description
SELK Polyclonal antibody specifically detects SELK in Human samples. It is validated for Western Blot, Immunocytochemistry
Specifications
| SELK | |
| Polyclonal | |
| Western Blot, Immunocytochemistry | |
| HSPC297, MGC17057, selenoprotein K, SelK | |
| Produced in rabbits immunized with E. coli-derived Human SELK fragment | |
| 0.1 mL | |
| Innate Immunity | |
| 58515 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunocytochemistry | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction