missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SELL, Mouse anti-Human, PE, Clone: HI62L, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB15353.100Reactions
This item is not returnable.
View return policy
Specifications
| SELL | |
| Monoclonal | |
| PE | |
| In PBS, pH 7.4 (protein stabilizer, 0.09% sodium azide). | |
| CD62L/LAM-1/LAM1/LECAM1/LNHR/LSEL/LYAM1/Leu-8/Lyam-1/PLNHR/TQ1/hLHRc | |
| Mouse | |
| 100 Reactions | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Flow Cytometry | |
| HI62L | |
| Flow Cytometry (20 uL/106 cells) The optimal working dilution should be determined by the end user. | |
| selectin L | |
| SELL | |
| Peripheral blood mononuclear cells. | |
| RUO | |
| 6402 | |
| Store in the dark at 4°C. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction