missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SELS Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32096C
This item is not returnable.
View return policy
Description
SELS Polyclonal antibody specifically detects SELS in Human samples. It is validated for Western Blot
Specifications
| SELS | |
| Polyclonal | |
| Western Blot | |
| ADO15, MGC104346, SBBI8, selenoprotein S, SelS, SEPS1, VCP-interacting membrane protein, VIMPMGC2553 | |
| The immunogen this antibody was made to, maps to a region between residue 125 to 175 of human Selenoprotein S using the numbering given in entry NP_982298.1 (GeneID 55829). | |
| 0.1 mL | |
| Signal Transduction | |
| 55829 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 650 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction