missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SELT Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA558927

Product Code. 15726801

  • € 566.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: CMMVFFLSNM IENQCMSTGA FEITLNDVPV WSKLESGHLP SMQQLVQILD NEMKLNVHMD SIPHHR Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

SELT encodes a selenoprotein, which contains a selenocysteine (Sec) residue at its active site. The selenocysteine is encoded by the UGA codon that normally signals translation termination. The 3' UTR of selenoprotein genes have a common stem-loop structure, the sec insertion sequence (SECIS), that is necessary for the recognition of UGA as a Sec codon rather than as a stop signal.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SELT
Polyclonal
Unconjugated
SELENOT
selenoprotein T; SELENOT; SELT; Thioredoxin reductase-like selenoprotein T; UNQ150/PRO176
Rabbit
Antigen Affinity Chromatography
RUO
51714
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
P62341
SELENOT
Recombinant protein corresponding to Human SELT. Recombinant protein control fragment (Product #RP-95735).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.