missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEMA3G Monoclonal Antibody (OTI2C3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies CF811391
This item is not returnable.
View return policy
Description
SEMA3G Monoclonal antibody specifically detects SEMA3G in Human samples. It is validated for Western BlotSpecifications
| SEMA3G | |
| Monoclonal | |
| See Label | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q9NS25 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 728695 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI2C3 | |
| Unconjugated | |
| SPANXB1 | |
| sem2; SEMA3G; semaphorin 3G; Semaphorin sem2; Semaphorin-3G | |
| Full length human recombinant protein of human SPANXB1 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Lyophilized |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction