missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEMA3G Monoclonal Antibody (OTI8B9), TrueMAB™, OriGene
Description
SEMA3G Monoclonal antibody specifically detects SEMA3G in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | SEMA3G |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | OTI8B9 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| Gene | SEMA3G |
| Gene Accession No. | Q9NS98 |
| Gene Alias | sem2; SEMA3G; semaphorin 3G; Semaphorin sem2; Semaphorin-3G |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?