Learn More
SEMA3G Mouse anti-Human, Clone: OTI8B10, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF811393
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Has chemorepulsive activities for sympathetic axons. Ligand of NRP2.Specifications
| SEMA3G | |
| Monoclonal | |
| Unconjugated | |
| SEMA3G | |
| sem2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 56920 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI8B10 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q9NS98 | |
| SEMA3G | |
| Human recombit protein fragment corresponding to amino acids 569-782 of human SEMA3G produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.