missing translation for 'onlineSavingsMsg'
Learn More

SEMA4D Monoclonal Antibody (OTI8G5), TrueMAB™, OriGene

Product Code. p-200015082
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17522454 30 μL 30µL
17502454 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17522454 Supplier Origene Technologies Supplier No. TA809851S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SEMA4D Monoclonal antibody specifically detects SEMA4D in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen SEMA4D
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI8G5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Sema4d
Gene Accession No. Q92854
Gene Alias A8; BB18; C9orf164; CD100; coll-4; GR3; M-Sema G; M-sema-G; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4D; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, 4D; sema J; Sema4d; Semacl2; SEMAJ; semaphorin 4D; semaphorin H; semaphorin-4D; Semaphorin-C-like 2; semaphorin-J; Semcl2
Host Species Mouse
Immunogen Full length human recombinant protein of human SEMA4D produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10507
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.