missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SEMA5A Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA531283
This item is not returnable.
View return policy
Description
Recommended positive controls: Mouse brain. Predicted reactivity: Mouse (95%), Rat (94%), Rhesus Monkey (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Members of the semaphorin protein family, such as SEMA5A, are involved in axonal guidance during neural development (Adams et al., 1996 [PubMed 8817451]).
Specifications
| SEMA5A | |
| Polyclonal | |
| Unconjugated | |
| SEMA5A | |
| 5930434A13; 9130201M22Rik; AI464145; Ensembl:ENSRNOP00000016506}; M-Sema D; sema domain, seven thrombospondin repeats (type 1 and type 1-like), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 5A; sema F; SEMA5A; sema5a {ECO:0000312; SEMAF; semaphorin 5A; semaphorin F; semaphorin-5A; semaphorin-5A {ECO:0000250; Semaphorin-F; semF; UniProtKB:Q13591} | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 20356, 9037 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q13591, Q62217 | |
| SEMA5A | |
| Recombinant protein encompassing a sequence within the Extracellular domain of human Semaphorin 5A. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction