Learn More
Invitrogen™ SEMA6A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5143762
Description
The antibody detects a 120kDa* protein corresponding to the apparent molecular mass of Sema-6A on SDS-PAGE immunoblots of Sema-6A transfected COS7 cells, human colorectal cancer cells, and rat PC12 cells. In addition, shRNA knockdown of Sema-6A expression significantly reduces detection of Sema-6A using anti-Sema-6A (C-terminus) antibody.
SEMA6A can act as repulsive axon guidance cues. It may play a role in channeling sympathetic axons into the sympathetic chains and controlling the temporal sequence of sympathetic target innervation.
Specifications
| SEMA6A | |
| Polyclonal | |
| Unconjugated | |
| SEMA6A | |
| 9330158E07; A730020P05Rik; AI851735; b2b997Clo; HT018; KIAA1368; Mutant line 997; SEMA; sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6A; sema Q; Sema VIA; SEMA6A; SEMA6A1; SEMA6A-1; semaphorin 6A; semaphorin 6A-1; semaphorin Q; Semaphorin VIA; semaphorin-6A; Semaphorin-6A-1; Semaq; VIA | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 20358, 361324, 57556 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide | |
| O35464, Q9H2E6 | |
| SEMA6A | |
| Semaphorin 6A synthetic peptide (coupled to carrier protein) corresponding to amino acids in the C-terminus of human Sema-6A. The sequence used is identical to similar regions in rat and mouse Sema-6A, and has low homology to other semaphorin family members. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.