missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SEMA6C Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA531199

Product Code. 13425717

  • € 530.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Recommended positive controls: NT2D1, IMR32, U87-MG, MCF-7. Predicted reactivity: Mouse (96%), Rat (96%), Rabbit (97%), Rhesus Monkey (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene product is a member of the semaphorin family of proteins. Semaphorins represent important molecular signals controlling multiple aspects of the cellular response that follows CNS injury, and thus may play an important role in neural regeneration.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SEMA6C
Polyclonal
Unconjugated
SEMA6C
KIAA1869; mKIAA1869; m-Sema Y; m-SemaY; m-SemaY2; sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6C; sema Y; SEMA6C; semaphorin 6c; semaphorin-6C; Semaphorin-Y; Semay
Rabbit
Antigen affinity chromatography
RUO
10500
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Neutralization, Western Blot
1 mg/mL
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Q9H3T2
SEMA6C
Recombinant protein encompassing a sequence within the Extracellular domain of human SEMA6C. The exact sequence is proprietary.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.