Learn More
Semadeni™ Semadeni Polypropylene Cup
Click to view available options
Capacity (Metric):
1100 mL
Unit Size:
Pack of 100
Specifications
Specifications
| Quantity | 100 Pk. |
| Capacity (Metric) | 1100 mL |
| Product Type | Storage Cup |
| Material | Polypropylene |
| For Use With (Application) | Suitable for food packaging. |
| Height (Metric) | 135 mm |