missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Semaphorin 4A Antibody (741531), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB4694J
This item is not returnable.
View return policy
Description
Semaphorin 4A Monoclonal antibody specifically detects Semaphorin 4A in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| Semaphorin 4A | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 741531 | |
| Flow Cytometry, CyTOF-ready | |
| CORD10, RP35, Sema B, sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, (semaphorin) 4A, SEMAB, semaphorin-4A, Semaphorin-B, SemB, SEMBFLJ12287 | |
| Mouse myeloma cell line NS0-derived recombinant human Semaphorin 4A, Gly32-His683, Accession # Q9H3S1 | |
| 0.1 mL | |
| Neuroscience, Vision | |
| 64218 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction